4-(4-Fluorophenyl)-3H,4H,5H,6H,7H-imidazo[4,5-c]pyridine 95%

Lieferant: Apollo Scientific

PC28012-1G
APOSPC28012-1GEA 671 EUR
APOSPC28012-1G
4-(4-Fluorophenyl)-3H,4H,5H,6H,7H-imidazo[4,5-c]pyridine 95%
4-(4-Fluorophenyl)-3H,4H,5H,6H,7H-imidazo[4,5-c]pyridine
Formel: C₁₂H₁₂FN₃
Lagertemperatur: Raumtemperatur
MDL: MFCD02571314
CAS-Nummer: 7271-09-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR