4-[3-(Trifluoromethyl)phenyl]-3H,4H,5H,6H,7H-imidazo[4,5-c]pyridine, tech

Lieferant: Apollo Scientific

PC28013-1G
APOSPC28013-1GEA 1570 EUR
APOSPC28013-1G
4-[3-(Trifluoromethyl)phenyl]-3H,4H,5H,6H,7H-imidazo[4,5-c]pyridine, tech
4-[3-(Trifluoromethyl)phenyl]-3H,4H,5H,6H,7H-imidazo[4,5-c]pyridine
Formel: C₁₃H₁₂F₃N₃
Lagertemperatur: Raumtemperatur
MDL: MFCD02102434
CAS-Nummer: 477871-72-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR