N-(2,4-Difluorophenyl)-2-(1-methyl-1H-pyrrol-2-yl)-2-oxoacetamide, tech

Lieferant: Apollo Scientific

PC28015-1G
APOSPC28015-1GEA 1570 EUR
APOSPC28015-1G
N-(2,4-Difluorophenyl)-2-(1-methyl-1H-pyrrol-2-yl)-2-oxoacetamide, tech
N-(2,4-Difluorophenyl)-2-(1-methyl-1H-pyrrol-2-yl)-2-oxoacetamide
Formel: C₁₃H₁₀F₂N₂O₂
Molecular Weight: 264,228 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD02102459
CAS-Nummer: 477872-05-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR