5-Methoxy-2-[3-(trifluoromethyl)phenyl]pyrimidin-4-ol, tech

Lieferant: Apollo Scientific

PC28016-1G
APOSPC28016-1GEA 0 EUR
APOSPC28016-1G
5-Methoxy-2-[3-(trifluoromethyl)phenyl]pyrimidin-4-ol, tech
5-Methoxy-2-[3-(trifluoromethyl)phenyl]pyrimidin-4-ol
Formel: C₁₂H₉F₃N₂O₂
Lagertemperatur: Raumtemperatur
MDL: MFCD04124708
CAS-Nummer: 860788-30-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR