Ethyl 2-[2-(trifluoromethyl)-1H-1,3-benzodiazol-1-yl]acetate 95%

Lieferant: Apollo Scientific

PC28017-1G
APOSPC28017-1GEA 671 EUR
APOSPC28017-1G
Ethyl 2-[2-(trifluoromethyl)-1H-1,3-benzodiazol-1-yl]acetate 95%
Ethyl 2-[2-(trifluoromethyl)-1H-1,3-benzodiazol-1-yl]acetate
Formel: C₁₂H₁₁F₃N₂O₂
Molecular Weight: 272,223 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD01956277
CAS-Nummer: 82791-95-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR