4-(Chlorodifluoromethyl)-2-hydroxy-6-methylpyridine-3-carbonitrile, tech
Lieferant: Apollo Scientific
PC28018-1G
APOSPC28018-1GEA
1570
EUR
APOSPC28018-1G
4-(Chlorodifluoromethyl)-2-hydroxy-6-methylpyridine-3-carbonitrile, tech
4-(Chlorodifluoromethyl)-2-hydroxy-6-methylpyridine-3-carbonitrile
Formel:
C₈H₅ClF₂N₂O Molecular Weight: 218,588 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD03011806 CAS-Nummer: 383147-78-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...