3-(2-Fluorophenyl)-3-(1-oxo-2,3-dihydro-1H-isoindol-2-yl)propanoic acid, tech
Lieferant: Apollo Scientific
PC28019-1G
APOSPC28019-1GEA
1570
EUR
APOSPC28019-1G
3-(2-Fluorophenyl)-3-(1-oxo-2,3-dihydro-1H-isoindol-2-yl)propanoic acid, tech
3-(2-Fluorophenyl)-3-(1-oxo-2,3-dihydro-1H-isoindol-2-yl)propanoic acid
Formel:
C₁₇H₁₄FNO₃ Molecular Weight: 299,296 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD03011837 CAS-Nummer: 383148-31-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...