5-Methoxy-2-(trifluoromethyl)benzonitrile, tech

Lieferant: Apollo Scientific

PC28020-1G
APOSPC28020-1GEA 1570 EUR
APOSPC28020-1G
5-Methoxy-2-(trifluoromethyl)benzonitrile, tech
5-Methoxy-2-(trifluoromethyl)benzonitrile
Formel: C₉H₆F₃NO
Molecular Weight: 201,145 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD13185686
CAS-Nummer: 1214346-63-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR