{2-[3-Chloro-5-(trifluoromethyl)pyridin-2-yl]ethyl}(methoxy)amine, tech

Lieferant: Apollo Scientific

PC28021-1G
APOSPC28021-1GEA 1570 EUR
APOSPC28021-1G
{2-[3-Chloro-5-(trifluoromethyl)pyridin-2-yl]ethyl}(methoxy)amine, tech
{2-[3-Chloro-5-(trifluoromethyl)pyridin-2-yl]ethyl}(methoxy)amine
Formel: C₉H₁₀ClF₃N₂O
Molecular Weight: 254,637 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD02571791
CAS-Nummer: 400078-95-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR