2-Amino-3-chloro-5-(trifluoromethyl)pyridin-1-ium-1-olate, tech

Lieferant: Apollo Scientific

PC28022-1G
APOSPC28022-1GEA 1570 EUR
APOSPC28022-1G
2-Amino-3-chloro-5-(trifluoromethyl)pyridin-1-ium-1-olate, tech
2-Amino-3-chloro-5-(trifluoromethyl)pyridin-1-ium-1-olate
Formel: C₆H₄ClF₃N₂O
Molecular Weight: 212,557 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00215312
CAS-Nummer: 338397-85-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR