2-[3-(Trifluoromethyl)phenoxy]ethan-1-amine hydrochloride 95%

Lieferant: Apollo Scientific

PC28023-1G
APOSPC28023-1GEA 306 EUR
APOSPC28023-1G
2-[3-(Trifluoromethyl)phenoxy]ethan-1-amine hydrochloride 95%
2-[3-(Trifluoromethyl)phenoxy]ethan-1-amine hydrochloride
Formel: C₉H₁₀F₃NO·ClH
Lagertemperatur: Raumtemperatur
MDL: MFCD00793842
CAS-Nummer: 104774-96-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR