3-[(2-Aminophenyl)amino]-5-(4-fluorophenyl)cyclohex-2-en-1-one, tech

Lieferant: Apollo Scientific

PC28026-1G
APOSPC28026-1GEA 1570 EUR
APOSPC28026-1G
3-[(2-Aminophenyl)amino]-5-(4-fluorophenyl)cyclohex-2-en-1-one, tech
3-[(2-Aminophenyl)amino]-5-(4-fluorophenyl)cyclohex-2-en-1-one
Formel: C₁₈H₁₇FN₂O
Molecular Weight: 296,339 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00794542
CAS-Nummer: 338400-92-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR