(3Z)-4-(Cyclohexylamino)-1,1,1-trifluoropent-3-en-2-one, tech
Lieferant: Apollo Scientific
PC28027-1G
APOSPC28027-1GEA
1570
EUR
APOSPC28027-1G
(3Z)-4-(Cyclohexylamino)-1,1,1-trifluoropent-3-en-2-one, tech
(3Z)-4-(Cyclohexylamino)-1,1,1-trifluoropent-3-en-2-one
Formel:
C₁₁H₁₆F₃NO Molecular Weight: 235,246 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD01315403 CAS-Nummer: 338401-97-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...