3-(1,1-Difluorobut-3-en-1-yl)-8-(trifluoromethyl)-[1,2,4]triazolo[4,3-a]pyridine, tech

Lieferant: Apollo Scientific

PC28028-1G
APOSPC28028-1GEA 1570 EUR
APOSPC28028-1G
3-(1,1-Difluorobut-3-en-1-yl)-8-(trifluoromethyl)-[1,2,4]triazolo[4,3-a]pyridine, tech
3-(1,1-Difluorobut-3-en-1-yl)-8-(trifluoromethyl)-[1,2,4]triazolo[4,3-a]pyridine
Formel: C₁₁H₈F₅N₃
Molecular Weight: 277,193 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD02571332
CAS-Nummer: 477889-55-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR