4-[(2-Chloro-4-fluorophenyl)methoxy]-1lambda6-thiomorpholine-1,1-dione, tech

Lieferant: Apollo Scientific

PC28029-1G
APOSPC28029-1GEA 1570 EUR
APOSPC28029-1G
4-[(2-Chloro-4-fluorophenyl)methoxy]-1lambda6-thiomorpholine-1,1-dione, tech
4-[(2-Chloro-4-fluorophenyl)methoxy]-1lambda6-thiomorpholine-1,1-dione
Formel: C₁₁H₁₃ClFNO₃S
Molecular Weight: 293,742 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD02102594
CAS-Nummer: 477889-61-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR