6-Chloro-3-(difluoromethyl)-[1,2,4]triazolo[4,3-b]pyridazine 95%
Lieferant: Apollo Scientific
PC28030-1G
APOSPC28030-1GEA
1570
EUR
APOSPC28030-1G
6-Chloro-3-(difluoromethyl)-[1,2,4]triazolo[4,3-b]pyridazine 95%
6-Chloro-3-(difluoromethyl)-[1,2,4]triazolo[4,3-b]pyridazine
Formel:
C₆H₃ClF₂N₄ Molecular Weight: 204,565 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD02186392 CAS-Nummer: 477889-88-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...