3-[(3-Fluorophenyl)methyl]-2,4-dimethylimidazo[1,5-a]pyrimidine-8-carbonitrile, tech

Lieferant: Apollo Scientific

PC28031-1G
APOSPC28031-1GEA 1570 EUR
APOSPC28031-1G
3-[(3-Fluorophenyl)methyl]-2,4-dimethylimidazo[1,5-a]pyrimidine-8-carbonitrile, tech
3-[(3-Fluorophenyl)methyl]-2,4-dimethylimidazo[1,5-a]pyrimidine-8-carbonitrile
Formel: C₁₆H₁₃FN₄
Molecular Weight: 280,3 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD02571880
CAS-Nummer: 477890-40-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR