Methyl 2-[2-(trifluoromethyl)-1H-1,3-benzodiazol-1-yl]acetate, tech
Lieferant: Apollo Scientific
PC28032-1G
APOSPC28032-1GEA
1570
EUR
APOSPC28032-1G
Methyl 2-[2-(trifluoromethyl)-1H-1,3-benzodiazol-1-yl]acetate, tech
Methyl 2-[2-(trifluoromethyl)-1H-1,3-benzodiazol-1-yl]acetate
Formel:
C₁₁H₉F₃N₂O₂ Molecular Weight: 258.197 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD02102654 CAS-Nummer: 139591-08-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...