1-[2-(Trifluoromethyl)-1H-1,3-benzodiazol-1-yl]propan-2-one, tech
Lieferant: Apollo Scientific
PC28033-1G
APOSPC28033-1GEA
1570
EUR
APOSPC28033-1G
1-[2-(Trifluoromethyl)-1H-1,3-benzodiazol-1-yl]propan-2-one, tech
1-[2-(Trifluoromethyl)-1H-1,3-benzodiazol-1-yl]propan-2-one
Formel:
C₁₁H₉F₃N₂O Molecular Weight: 242,197 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD01419489 CAS-Nummer: 321980-88-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...