2-[2-(Trifluoromethyl)-1H-1,3-benzodiazol-1-yl]propanoic acid, tech

Lieferant: Apollo Scientific

PC28034-1G
APOSPC28034-1GEA 1570 EUR
APOSPC28034-1G
2-[2-(Trifluoromethyl)-1H-1,3-benzodiazol-1-yl]propanoic acid, tech
2-[2-(Trifluoromethyl)-1H-1,3-benzodiazol-1-yl]propanoic acid
Formel: C₁₁H₉F₃N₂O₂
Molecular Weight: 258,197 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD02571586
CAS-Nummer: 478030-59-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR