2,2,2-Trifluoro-N-(4-methoxy-3-oxo-2,3-dihydro-1H-inden-1-yl)acetamide, tech

Lieferant: Apollo Scientific

PC28035-1G
APOSPC28035-1GEA 1570 EUR
APOSPC28035-1G
2,2,2-Trifluoro-N-(4-methoxy-3-oxo-2,3-dihydro-1H-inden-1-yl)acetamide, tech
2,2,2-Trifluoro-N-(4-methoxy-3-oxo-2,3-dihydro-1H-inden-1-yl)acetamide
Formel: C₁₂H₁₀F₃NO₃
Molecular Weight: 273,208 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD03102804
CAS-Nummer: 861209-50-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR