(2E)-1-[3-Chloro-5-(trifluoromethyl)pyridin-2-yl]-3-(dimethylamino)prop-2-en-1-one, tech

Lieferant: Apollo Scientific

PC28036-1G
APOSPC28036-1GEA 1570 EUR
APOSPC28036-1G
(2E)-1-[3-Chloro-5-(trifluoromethyl)pyridin-2-yl]-3-(dimethylamino)prop-2-en-1-one, tech
(2E)-1-[3-Chloro-5-(trifluoromethyl)pyridin-2-yl]-3-(dimethylamino)prop-2-en-1-one
Formel: C₁₁H₁₀ClF₃N₂O
Molecular Weight: 278,658 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD04124942
CAS-Nummer: 1107035-71-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR