4,4,4-Trifluoro-1-(2-methoxyphenyl)butane-1,3-dione 95%

Lieferant: Apollo Scientific

PC28037-1G
APOSPC28037-1GEA 427 EUR
APOSPC28037-1G
4,4,4-Trifluoro-1-(2-methoxyphenyl)butane-1,3-dione 95%
4,4,4-Trifluoro-1-(2-methoxyphenyl)butane-1,3-dione
Formel: C₁₁H₉F₃O₃
Molecular Weight: 246,183 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD03419768
CAS-Nummer: 15191-69-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR