3-[3-Fluoro-4-(morpholin-4-yl)phenyl]-3-oxopropanenitrile, tech

Lieferant: Apollo Scientific

PC28039-1G
APOSPC28039-1GEA 1570 EUR
APOSPC28039-1G
3-[3-Fluoro-4-(morpholin-4-yl)phenyl]-3-oxopropanenitrile, tech
3-[3-Fluoro-4-(morpholin-4-yl)phenyl]-3-oxopropanenitrile
Formel: C₁₃H₁₃FN₂O₂
Molecular Weight: 248,253 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD06496173
CAS-Nummer: 861211-70-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR