4-Fluorophenyl (2E)-3-(dimethylamino)prop-2-enoate, tech

Lieferant: Apollo Scientific

PC28041-1G
APOSPC28041-1GEA 1570 EUR
APOSPC28041-1G
4-Fluorophenyl (2E)-3-(dimethylamino)prop-2-enoate, tech
4-Fluorophenyl (2E)-3-(dimethylamino)prop-2-enoate
Formel: C₁₁H₁₂FNO₂
Molecular Weight: 209,217 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00201970
CAS-Nummer: 338404-21-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR