Methyl 3-hydroxy-3-(trifluoromethyl)-hexahydro-2H-[1,2]oxazolo[2,3-a]pyridine-2-carboxylate, tech

Lieferant: Apollo Scientific

PC28043-1G
APOSPC28043-1GEA 1570 EUR
APOSPC28043-1G
Methyl 3-hydroxy-3-(trifluoromethyl)-hexahydro-2H-[1,2]oxazolo[2,3-a]pyridine-2-carboxylate, tech
Methyl 3-hydroxy-3-(trifluoromethyl)-hexahydro-2H-[1,2]oxazolo[2,3-a]pyridine-2-carboxylate
Formel: C₁₀H₁₄F₃NO₄
Molecular Weight: 269,218 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00172848
CAS-Nummer: 317833-14-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR