7-[4-(3-Fluoropropoxy)phenyl]-[1,2,4]triazolo[1,5-a]pyrimidine, tech

Lieferant: Apollo Scientific

PC28046-1G
APOSPC28046-1GEA 1570 EUR
APOSPC28046-1G
7-[4-(3-Fluoropropoxy)phenyl]-[1,2,4]triazolo[1,5-a]pyrimidine, tech
7-[4-(3-Fluoropropoxy)phenyl]-[1,2,4]triazolo[1,5-a]pyrimidine
Formel: C₁₄H₁₃FN₄O
Molecular Weight: 272,278 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD02571714
CAS-Nummer: 478039-24-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR