7-[4-(3-Fluoropropoxy)phenyl]-2-methylpyrazolo[1,5-a]pyrimidine, tech

Lieferant: Apollo Scientific

PC28047-1G
APOSPC28047-1GEA 1570 EUR
APOSPC28047-1G
7-[4-(3-Fluoropropoxy)phenyl]-2-methylpyrazolo[1,5-a]pyrimidine, tech
7-[4-(3-Fluoropropoxy)phenyl]-2-methylpyrazolo[1,5-a]pyrimidine
Formel: C₁₆H₁₆FN₃O
Molecular Weight: 285,316 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD02571885
CAS-Nummer: 478039-30-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR