2,6-Difluoro-N-(1-hydroxy-3-phenylpropan-2-yl)benzamide, tech

Lieferant: Apollo Scientific

PC28048-1G
APOSPC28048-1GEA 1570 EUR
APOSPC28048-1G
2,6-Difluoro-N-(1-hydroxy-3-phenylpropan-2-yl)benzamide, tech
2,6-Difluoro-N-(1-hydroxy-3-phenylpropan-2-yl)benzamide
Formel: C₁₆H₁₅F₂NO₂
Molecular Weight: 291,293 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD02186157
CAS-Nummer: 478040-51-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR