(3E)-1,1,1-Trifluoro-4-[(pyridin-3-yl)amino]but-3-en-2-one, tech
Lieferant: Apollo Scientific
PC28049-1G
APOSPC28049-1GEA
1570
EUR
APOSPC28049-1G
(3E)-1,1,1-Trifluoro-4-[(pyridin-3-yl)amino]but-3-en-2-one, tech
(3E)-1,1,1-Trifluoro-4-[(pyridin-3-yl)amino]but-3-en-2-one
Formel:
C₉H₇F₃N₂O Molecular Weight: 216,16 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD01972158 CAS-Nummer: 428842-24-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...