(3E)-4-(2,3-Dihydro-1H-indol-1-yl)-1,1,1-trifluorobut-3-en-2-one, tech
Lieferant: Apollo Scientific
PC28050-1G
APOSPC28050-1GEA
1570
EUR
APOSPC28050-1G
(3E)-4-(2,3-Dihydro-1H-indol-1-yl)-1,1,1-trifluorobut-3-en-2-one, tech
(3E)-4-(2,3-Dihydro-1H-indol-1-yl)-1,1,1-trifluorobut-3-en-2-one
Formel:
C₁₂H₁₀F₃NO Molecular Weight: 241,209 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD02186219 CAS-Nummer: 428842-30-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...