(3Z)-4-(2,3-Dihydro-1H-indol-1-yl)-1,1-difluorobut-3-en-2-one, tech

Lieferant: Apollo Scientific

PC28051-1G
APOSPC28051-1GEA 1570 EUR
APOSPC28051-1G
(3Z)-4-(2,3-Dihydro-1H-indol-1-yl)-1,1-difluorobut-3-en-2-one, tech
(3Z)-4-(2,3-Dihydro-1H-indol-1-yl)-1,1-difluorobut-3-en-2-one
Formel: C₁₂H₁₁F₂NO
Molecular Weight: 223,219 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD02186227
CAS-Nummer: 478040-92-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR