2-{3-[(4-Fluorophenyl)methyl]-4-oxo-3,4-dihydrophthalazin-1-yl}acetonitrile, tech

Lieferant: Apollo Scientific

PC28053-1G
APOSPC28053-1GEA 1570 EUR
APOSPC28053-1G
2-{3-[(4-Fluorophenyl)methyl]-4-oxo-3,4-dihydrophthalazin-1-yl}acetonitrile, tech
2-{3-[(4-Fluorophenyl)methyl]-4-oxo-3,4-dihydrophthalazin-1-yl}acetonitrile
Formel: C₁₇H₁₂FN₃O
Molecular Weight: 293,295 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD02186453
CAS-Nummer: 478041-35-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR