2-[2-(Trifluoromethyl)-1H-1,3-benzodiazol-1-yl]acetonitrile 95%

Lieferant: Apollo Scientific

PC28054-1G
APOSPC28054-1GEA 671 EUR
APOSPC28054-1G
2-[2-(Trifluoromethyl)-1H-1,3-benzodiazol-1-yl]acetonitrile 95%
2-[2-(Trifluoromethyl)-1H-1,3-benzodiazol-1-yl]acetonitrile
Formel: C₁₀H₆F₃N₃
Molecular Weight: 225,17 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD02187083
CAS-Nummer: 478042-43-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR