3-[3-Chloro-5-(trifluoromethyl)pyridin-2-yl]prop-2-yn-1-ol, tech

Lieferant: Apollo Scientific

PC28056-1G
APOSPC28056-1GEA 1570 EUR
APOSPC28056-1G
3-[3-Chloro-5-(trifluoromethyl)pyridin-2-yl]prop-2-yn-1-ol, tech
3-[3-Chloro-5-(trifluoromethyl)pyridin-2-yl]prop-2-yn-1-ol
Formel: C₉H₅ClF₃NO
Molecular Weight: 235,59 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD04125169
CAS-Nummer: 865657-82-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR