3-(4-Fluorophenyl)-5-methylpyrazolo[1,5-a]pyrimidin-7-amine, tech

Lieferant: Apollo Scientific

PC28059-1G
APOSPC28059-1GEA 1570 EUR
APOSPC28059-1G
3-(4-Fluorophenyl)-5-methylpyrazolo[1,5-a]pyrimidin-7-amine, tech
3-(4-Fluorophenyl)-5-methylpyrazolo[1,5-a]pyrimidin-7-amine
Formel: C₁₃H₁₁FN₄
Molecular Weight: 242,252 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD03617323
CAS-Nummer: 685106-57-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR