3-(4-Chloro-2-fluorophenyl)prop-2-yn-1-ol 95%

Lieferant: Apollo Scientific

PC28062-1G
APOSPC28062-1GEA 1330 EUR
APOSPC28062-1G
3-(4-Chloro-2-fluorophenyl)prop-2-yn-1-ol 95%
3-(4-Chloro-2-fluorophenyl)prop-2-yn-1-ol
Formel: C₉H₆ClFO
Lagertemperatur: Raumtemperatur
MDL: MFCD03787115
CAS-Nummer: 865659-21-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR