Ethyl 1-(3-fluorophenyl)-3,5-dimethyl-1H-pyrazole-4-carboxylate, tech
Lieferant: Apollo Scientific
PC28063-1G
APOSPC28063-1GEA
1570
EUR
APOSPC28063-1G
Ethyl 1-(3-fluorophenyl)-3,5-dimethyl-1H-pyrazole-4-carboxylate, tech
Ethyl 1-(3-fluorophenyl)-3,5-dimethyl-1H-pyrazole-4-carboxylate
Formel:
C₁₄H₁₅FN₂O₂ Molecular Weight: 262,28 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD07021394 CAS-Nummer: 346441-17-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...