N-Methyl-2-(pyridin-2-yl)-6-(trifluoromethyl)pyrimidin-4-amine, tech

Lieferant: Apollo Scientific

PC28064-1G
APOSPC28064-1GEA 1570 EUR
APOSPC28064-1G
N-Methyl-2-(pyridin-2-yl)-6-(trifluoromethyl)pyrimidin-4-amine, tech
N-Methyl-2-(pyridin-2-yl)-6-(trifluoromethyl)pyrimidin-4-amine
Formel: C₁₁H₉F₃N₄
Molecular Weight: 254,211 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00139797
CAS-Nummer: 338418-10-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR