1-Chloro-3-[4-(2-fluorophenyl)piperazin-1-yl]propan-2-ol, tech
Lieferant: Apollo Scientific
PC28065-1G
APOSPC28065-1GEA
0
EUR
APOSPC28065-1G
1-Chloro-3-[4-(2-fluorophenyl)piperazin-1-yl]propan-2-ol, tech
1-Chloro-3-[4-(2-fluorophenyl)piperazin-1-yl]propan-2-ol
Formel:
C₁₃H₁₈ClFN₂O Lagertemperatur: Raumtemperatur |
MDL:
MFCD04125368 CAS-Nummer: 865660-68-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...