3-[3-Chloro-5-(trifluoromethyl)pyridin-2-yl]propanoic acid, tech

Lieferant: Apollo Scientific

PC28067-1G
APOSPC28067-1GEA 1570 EUR
APOSPC28067-1G
3-[3-Chloro-5-(trifluoromethyl)pyridin-2-yl]propanoic acid, tech
3-[3-Chloro-5-(trifluoromethyl)pyridin-2-yl]propanoic acid
Formel: C₉H₇ClF₃NO₂
Molecular Weight: 253,606 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00794560
CAS-Nummer: 338422-79-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR