(2Z)-3-(4-Fluorophenyl)-2-[(1E)-(methoxyimino)methyl]prop-2-enenitrile, tech

Lieferant: Apollo Scientific

PC28068-1G
APOSPC28068-1GEA 1570 EUR
APOSPC28068-1G
(2Z)-3-(4-Fluorophenyl)-2-[(1E)-(methoxyimino)methyl]prop-2-enenitrile, tech
(2Z)-3-(4-Fluorophenyl)-2-[(1E)-(methoxyimino)methyl]prop-2-enenitrile
Formel: C₁₁H₉FN₂O
Molecular Weight: 204,2 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00663699
CAS-Nummer: 339278-28-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR