3-(Benzylamino)-1,1,1-trifluoropropan-2-ol 95%

Lieferant: Apollo Scientific

PC28071-1G
APOSPC28071-1GEA 1570 EUR
APOSPC28071-1G
3-(Benzylamino)-1,1,1-trifluoropropan-2-ol 95%
3-(Benzylamino)-1,1,1-trifluoropropan-2-ol
Formel: C₁₀H₁₂F₃NO
Molecular Weight: 219,204 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00551089
CAS-Nummer: 178218-36-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR