(3E)-4-(Benzylamino)-1,1,1-trifluorobut-3-en-2-one, tech
Lieferant: Apollo Scientific
PC28074-1G
APOSPC28074-1GEA
1570
EUR
APOSPC28074-1G
(3E)-4-(Benzylamino)-1,1,1-trifluorobut-3-en-2-one, tech
(3E)-4-(Benzylamino)-1,1,1-trifluorobut-3-en-2-one
Formel:
C₁₁H₁₀F₃NO Molecular Weight: 229,198 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD01931068 CAS-Nummer: 134219-70-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...