(3E)-4-(4-Benzylpiperazin-1-yl)-1,1,1-trifluorobut-3-en-2-one, tech
Lieferant: Apollo Scientific
PC28075-1G
APOSPC28075-1GEA
1570
EUR
APOSPC28075-1G
(3E)-4-(4-Benzylpiperazin-1-yl)-1,1,1-trifluorobut-3-en-2-one, tech
(3E)-4-(4-Benzylpiperazin-1-yl)-1,1,1-trifluorobut-3-en-2-one
Formel:
C₁₅H₁₇F₃N₂O Molecular Weight: 298,303 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD02186481 CAS-Nummer: 478047-08-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...