1,1,1-Trifluoro-3-(pyridin-2-ylsulphanyl)propan-2-ol, tech

Lieferant: Apollo Scientific

PC28076-1G
APOSPC28076-1GEA 1570 EUR
APOSPC28076-1G
1,1,1-Trifluoro-3-(pyridin-2-ylsulphanyl)propan-2-ol, tech
1,1,1-Trifluoro-3-(pyridin-2-ylsulphanyl)propan-2-ol
Formel: C₈H₈F₃NOS
Lagertemperatur: Raumtemperatur
MDL: MFCD02570800
CAS-Nummer: 478047-11-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR