N-Methyl-3-phenyl-6-(trifluoromethyl)-1,2,4-triazin-5-amine, tech

Lieferant: Apollo Scientific

PC28077-1G
APOSPC28077-1GEA 1570 EUR
APOSPC28077-1G
N-Methyl-3-phenyl-6-(trifluoromethyl)-1,2,4-triazin-5-amine, tech
N-Methyl-3-phenyl-6-(trifluoromethyl)-1,2,4-triazin-5-amine
Formel: C₁₁H₉F₃N₄
Molecular Weight: 254,211 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD03012065
CAS-Nummer: 383148-46-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR