3-(2-Chloroethyl)-1-[3-(trifluoromethyl)phenyl]urea, tech
Lieferant: Apollo Scientific
|
Achtung
|
PC28078-500MG
APOSPC28078-500MGEA
0
EUR
APOSPC28078-500MG
3-(2-Chloroethyl)-1-[3-(trifluoromethyl)phenyl]urea, tech
3-(2-Chloroethyl)-1-[3-(trifluoromethyl)phenyl]urea
Formel:
C₁₀H₁₀ClF₃N₂O Lagertemperatur: Raumtemperatur |
MDL:
MFCD03787347 CAS-Nummer: 500546-66-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...