5-[3-(Trifluoromethyl)phenyl]pyrazolo[1,5-a]pyrimidine-3-carbonitrile, tech

Lieferant: Apollo Scientific

PC28079-1G
APOSPC28079-1GEA 1570 EUR
APOSPC28079-1G
5-[3-(Trifluoromethyl)phenyl]pyrazolo[1,5-a]pyrimidine-3-carbonitrile, tech
5-[3-(Trifluoromethyl)phenyl]pyrazolo[1,5-a]pyrimidine-3-carbonitrile
Formel: C₁₄H₇F₃N₄
Molecular Weight: 288,227 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD04125471
CAS-Nummer: 866019-49-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR