4-[3-(Trifluoromethyl)phenyl]pyrimidin-2-amine, tech

Lieferant: Apollo Scientific

PC28080-1G
APOSPC28080-1GEA 0 EUR
APOSPC28080-1G
4-[3-(Trifluoromethyl)phenyl]pyrimidin-2-amine, tech
4-[3-(Trifluoromethyl)phenyl]pyrimidin-2-amine
Formel: C₁₁H₈F₃N₃
Lagertemperatur: Raumtemperatur
MDL: MFCD03407835
CAS-Nummer: 866019-76-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR